Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Bromo-6-chlorohexane | 6294-17-3 | 99.18% | 199.52 | 50 G
SDP

Catalog No. 5000453106
Encompass_Preferred

1-Bromo-6-chlorohexane is a PROTAC linker, a chemical compound used in the synthesis of PROTACs. This liquid appears colorless to light yellow and has a purity of 99.18%.

  • Used as a PROTAC linker in synthesis.
  • High purity of 99.18%.
  • Molecular weight of 199.52.
  • Chemical formula is C6H12BrCl.
  • Appears as a colorless to light yellow liquid.
  • Causes skin and serious eye irritation.
  • May cause respiratory irritation.
  • Store pure form at -20°C for 3 years or 4°C for 2 years.

Catalog No. 50-004-53106 Supplier Medchemexpress LLC Supplier No. HYW00978750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Bromo-6-chlorohexane is a PROTAC linker, a chemical compound used in the synthesis of PROTACs. This liquid appears colorless to light yellow and has a purity of 99.18%.

  • Used as a PROTAC linker in synthesis.
  • High purity of 99.18%.
  • Molecular weight of 199.52.
  • Chemical formula is C6H12BrCl.
  • Appears as a colorless to light yellow liquid.
  • Causes skin and serious eye irritation.
  • May cause respiratory irritation.
  • Store pure form at -20°C for 3 years or 4°C for 2 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.