Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-7-chloroheptane 95.0+%, TCI America™
SDP

Catalog No. B35375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35375G Supplier TCI America Supplier No. B35375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68105-93-1
Molecular Formula C7H14BrCl
Molecular Weight (g/mol) 213.543
MDL Number MFCD00270371
InChI Key ZMAOWMBSLZYILL-UHFFFAOYSA-N
PubChem CID 11665662
IUPAC Name 1-bromo-7-chloroheptane
SMILES C(CCCCl)CCCBr

Specifications

Color Yellow
Boiling Point 118°C
Quantity 5 g
Formula Weight 213.54
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-7-chloroheptane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.