Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-8-iodonaphthalene 98.0+%, TCI America™
SDP

Catalog No. B52201G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52201G 1 g
B5220200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52201G Supplier TCI America Supplier No. B52201G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4044-58-0
Molecular Formula C10H6BrI
Molecular Weight (g/mol) 332.97
MDL Number MFCD11656276
InChI Key XNDLRPFISURYTA-UHFFFAOYSA-N
PubChem CID 12347526
IUPAC Name 1-bromo-8-iodonaphthalene
SMILES BrC1=C2C(I)=CC=CC2=CC=C1

Specifications

Melting Point 101°C
Boiling Point 168°C
Quantity 1 g
Formula Weight 332.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-8-iodonaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.