Learn More
Medchemexpress LLC 1-Bromoadamantane | 768-90-1 | 99.5% | 215.13 | 50 G
SDP
Catalog No. 5000452715
Encompass_Preferred
1-Bromoadamantane is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also an important intermediate used in synthesizing medicines and new materials etc.
  • Can be used as a biological material
  • Can be used as an organic compound for life science related research
  • Important intermediate in synthesizing medicines
  • Important intermediate in synthesizing new materials
Catalog No. 50-004-52715 Supplier Medchemexpress LLC Supplier No. HYW00859450G
1-Bromoadamantane is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also an important intermediate used in synthesizing medicines and new materials etc.
  • Can be used as a biological material
  • Can be used as an organic compound for life science related research
  • Important intermediate in synthesizing medicines
  • Important intermediate in synthesizing new materials