Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromoanthracene (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. B34751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34751G Supplier TCI America Supplier No. B34751G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7397-92-4
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD08276282
InChI Key XMWJLKOCNKJERQ-UHFFFAOYSA-N
PubChem CID 12529827
IUPAC Name 1-bromoanthracene
SMILES C1=CC=C2C=C3C(=CC2=C1)C=CC=C3Br

Specifications

Melting Point 100°C
Quantity 1 g
Formula Weight 257.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromoanthracene (purified by sublimation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.