Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Bromobutane

Catalog No. B5949725KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5949725KG 2.5 kg
B594971KG 1 kg
B5949725G 25 g
B59497500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. B5949725KG Supplier Sigma Aldrich Supplier No. B594972.5KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Bromobutane

Specifications

CAS 109-65-9
Melting Point -112°C
Density 1.276 g/mL at 25°C
Boiling Point 100°C to 104°C
Molecular Formula C4H9Br
Refractive Index n20/D 1.439
Linear Formula CH3(CH2)3Br
MDL Number MFCD00000260
Quantity 2.5 kg
Synonym Butyl bromide
Molecular Weight (g/mol) 137.02 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.