Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Bromodecane

Catalog No. 145785100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-578-5100G 100 g
14-578-52KG 2 kg
14-578-5500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-578-5100G Supplier Sigma Aldrich Supplier No. 145785100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-bromodecane participates in alkylation of pentaerythritol and introduces two lipophilic groups in pentaerythritol
  • It reacts with 1,2-dimethylimidazole to yield 1-decyl-2,3-dimethylimidazolium bromide.1-bromodecane was used in the synthesis of ferrocene containing hexacatenar metallomesogen

Specifications

CAS 112-29-8
Density 1.066 g/mL (at 25°C (literature))
Boiling Point 238°C (lit.)
Molecular Formula C10H21Br
Refractive Index n20/D 1.456 (literature)
Linear Formula CH3(CH2)9 Br
MDL Number MFCD00000221
Quantity 100 g
Synonym Decyl bromide
Molecular Weight (g/mol) 221.18
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.