Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Bromododecane | 143-15-7 | 99.17% | 249.24 | 50 G
SDP

Catalog No. 5000452652
Encompass_Preferred

1-Bromododecane is an organic compound that serves as a versatile reagent in organic synthesis. It typically presents as a colorless to light yellow liquid.

  • Molecular weight is 249.24
  • Chemical formula is C12H25Br
  • CAS number is 143-15-7
  • Typical appearance is a liquid
  • Color ranges from colorless to light yellow
  • Purity for this batch is 99.17%
  • Recommended storage at room temperature for up to 3 years
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year
  • Ships at room temperature within the continental US
  • Intended for research use only

Catalog No. 50-004-52652 Supplier Medchemexpress LLC Supplier No. HYW00875850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Bromododecane is an organic compound that serves as a versatile reagent in organic synthesis. It typically presents as a colorless to light yellow liquid.

  • Molecular weight is 249.24
  • Chemical formula is C12H25Br
  • CAS number is 143-15-7
  • Typical appearance is a liquid
  • Color ranges from colorless to light yellow
  • Purity for this batch is 99.17%
  • Recommended storage at room temperature for up to 3 years
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year
  • Ships at room temperature within the continental US
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.