Learn More
Medchemexpress LLC 1-Bromododecane-d25 | 204259-66-5 | 99.85% | 274.38 | 5 MG
SDP
Catalog No. 5000453390
Encompass_Preferred
1-Bromododecane-d25 is the deuterium labeled 1-Bromododecane. It is intended for research use only, and the company does not sell to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration is noted for its potential to impact the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-53390 Supplier Medchemexpress LLC Supplier No. HYW008758S5MG
1-Bromododecane-d25 is the deuterium labeled 1-Bromododecane. It is intended for research use only, and the company does not sell to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration is noted for its potential to impact the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.