Learn More
Medchemexpress LLC 1-Bromododecane-d25 | 204259-66-5 | 99.9% | 274.38 | 10 MG
SDP
Catalog No. 5000453077
Encompass_Preferred
1-Bromododecane-d25 is the deuterium labeled 1-Bromododecane. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suitable for research use only
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-53077 Supplier Medchemexpress LLC Supplier No. HYW008758S10MG
Add to Cart
Add to Cart
1-Bromododecane-d25 is the deuterium labeled 1-Bromododecane. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suitable for research use only
  • Potential to affect pharmacokinetic and metabolic profiles of drugs