Learn More
Medchemexpress LLC 1-Bromododecane-d3 | 204259-68-7 | 252.25 | 25 MG
SDP
Catalog No. 5000451481
Encompass_Preferred
1-Bromododecane-d3 is a deuterium labeled chemical primarily used in laboratory settings and for the manufacture of substances. It is a stable heavy isotope incorporated into drug molecules, serving as a tracer for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Suitable for laboratory chemical applications
  • Used in the manufacture of various substances
  • Incorporates stable heavy isotopes
  • Can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-51481 Supplier Medchemexpress LLC Supplier No. 25MGHYW008758S1
1-Bromododecane-d3 is a deuterium labeled chemical primarily used in laboratory settings and for the manufacture of substances. It is a stable heavy isotope incorporated into drug molecules, serving as a tracer for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Suitable for laboratory chemical applications
  • Used in the manufacture of various substances
  • Incorporates stable heavy isotopes
  • Can affect pharmacokinetic and metabolic profiles of drugs