Learn More
Medchemexpress LLC 1-Bromododecane-d3 | 204259-68-7 | 98.0% | 252.25 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW008758S15MG
Encompass_Preferred
1-Bromododecane-d3 is the deuterium labeled 1-Bromododecane. Deuteration has garnered attention due to its potential to impact the pharmacokinetic and metabolic profiles of drugs. Stable heavy isotopes of hydrogen, carbon, and other elements have been integrated into drug molecules primarily as tracers for quantitative analysis during the drug development process.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-51165