Learn More
Medchemexpress LLC (±)-(1-bromoethyl-2,2,2-d3)benzene | 23088-42-8 | 98.7% | 188.08 g·mol⁻¹ | C8H6D3Br | 5 MG
SDP
Catalog No. 5000302185
Encompass_Preferred
(±)-(1-Bromoethyl-2,2,2-d3)benzene (CAS 23088-42-8) is the deuterium-labeled analogue of (1-bromoethyl)benzene supplied as a light yellow to light brown liquid. It is intended for use as a stable-isotope internal standard and tracer in analytical and drug-development workflows, enabling accurate quantitation and tracking in NMR, GC-MS, and LC-MS applications.
  • Deuterium-labeled (d3) ethyl group provides a clear mass shift for MS analyses.
  • High purity (98.7%) suitable for analytical applications.
  • Chemical formula C8H6D3Br and molecular weight 188.08 g·mol⁻¹.
  • Liquid appearance; soluble in common organic solvents for easy sample preparation.
  • Effective internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Useful tracer for metabolic and pharmacokinetic studies.
Catalog No. 50-003-02185 Supplier Medchemexpress LLC Supplier No. HYW016254S5MG
(±)-(1-Bromoethyl-2,2,2-d3)benzene (CAS 23088-42-8) is the deuterium-labeled analogue of (1-bromoethyl)benzene supplied as a light yellow to light brown liquid. It is intended for use as a stable-isotope internal standard and tracer in analytical and drug-development workflows, enabling accurate quantitation and tracking in NMR, GC-MS, and LC-MS applications.
  • Deuterium-labeled (d3) ethyl group provides a clear mass shift for MS analyses.
  • High purity (98.7%) suitable for analytical applications.
  • Chemical formula C8H6D3Br and molecular weight 188.08 g·mol⁻¹.
  • Liquid appearance; soluble in common organic solvents for easy sample preparation.
  • Effective internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Useful tracer for metabolic and pharmacokinetic studies.