Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1-bromoethyl)benzene-d3 | 23088-42-8 | 98.7% | 188.08 | C8H6D3Br | 50 MG
SDP

Catalog No. 5000302181
Encompass_Preferred

(1-Bromoethyl)benzene-d3 is the deuterated analogue of (1-bromoethyl)benzene intended for research and analytical use. Provided as a light yellow to light brown liquid with high chemical purity and defined isotopic labeling, it is suitable as an internal standard for mass spectrometry, isotopic tracing, and analytical method development.

  • Deuterium labeled (d3) molecular analogue for isotopic tracing.
  • High purity, 98.7%.
  • CAS number 23088-42-8 and molecular weight 188.08.
  • Molecular formula C8H6D3Br.
  • Supplied as a liquid; color light yellow to light brown.
  • Pack sizes include 1 mg, 5 mg, 10 mg, 25 mg, 50 mg.
  • Intended uses include mass spectrometry internal standard and analytical reference.

Catalog No. 50-003-02181 Supplier Medchemexpress LLC Supplier No. HYW016254S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1-Bromoethyl)benzene-d3 is the deuterated analogue of (1-bromoethyl)benzene intended for research and analytical use. Provided as a light yellow to light brown liquid with high chemical purity and defined isotopic labeling, it is suitable as an internal standard for mass spectrometry, isotopic tracing, and analytical method development.

  • Deuterium labeled (d3) molecular analogue for isotopic tracing.
  • High purity, 98.7%.
  • CAS number 23088-42-8 and molecular weight 188.08.
  • Molecular formula C8H6D3Br.
  • Supplied as a liquid; color light yellow to light brown.
  • Pack sizes include 1 mg, 5 mg, 10 mg, 25 mg, 50 mg.
  • Intended uses include mass spectrometry internal standard and analytical reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.