Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Bromoethyl)benzene 95.0+%, TCI America™
SDP

Catalog No. B0592100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0592100G 100 g
B059225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0592100G Supplier TCI America Supplier No. B0592100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 585-71-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000139
InChI Key CRRUGYDDEMGVDY-UHFFFAOYSA-N
Synonym 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene
PubChem CID 11454
IUPAC Name 1-bromoethylbenzene
SMILES CC(C1=CC=CC=C1)Br

Specifications

Color Yellow
Boiling Point 200°C
Quantity 100 g
Formula Weight 185.06
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (1-Bromoethyl)benzene
EINECSNumber (3)-0035
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.