Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-bromoethylbenzene-d3 | 23088-42-8 | 98.7% | 188.08 | C8H6D3Br | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYW016254S

Encompass_Preferred

(1-Bromoethyl)benzene-d3 is the deuterium-labeled isotopologue of (1-Bromoethyl)benzene intended for research and analytical applications. It is supplied in small milligram quantities and is accompanied by documentation for safe handling and quality verification.

  • Deuterium-labeled isotopologue for use as a stable isotope standard or tracer.
  • Purity 98.7%.
  • Molecular formula C8H6D3, molecular weight 188.08.
  • Appearance: liquid, light yellow to light brown.
  • Available in small milligram packages for analytical workflows.
  • Data sheet, safety data sheet, and certificate of analysis available for download.

Catalog No. 50-003-03532


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.