Learn More
Medchemexpress LLC 1-bromoheptane-d15 | 98195-42-7 | 194.19 g·mol⁻¹ | C7D15Br | 50 MG
SDP
Catalog No. 5000300782
Encompass_Preferred
1-Bromoheptane-d15 is a deuterium-labeled analogue of 1-bromoheptane (C7D15Br, MW 194.19 g·mol⁻¹) used as an isotopic standard and reagent in synthetic and analytical chemistry. It is supplied as a liquid, typically colorless to light yellow, and is intended for applications that require deuterium incorporation or accurate mass-based quantitation.
  • Deuterium-labeled (d15) for isotopic tracing and mass-spectrometry quantitation
  • Chemical formula C7D15Br; molecular weight 194.19 g·mol⁻¹
  • Supplied as a liquid, colorless to light yellow
  • Suitable as an internal standard for mass spectrometry
  • Useful in mechanistic studies, tracer experiments, and synthetic applications requiring deuterium
Catalog No. 50-003-00782 Supplier Medchemexpress LLC Supplier No. HYW016577S50MG
1-Bromoheptane-d15 is a deuterium-labeled analogue of 1-bromoheptane (C7D15Br, MW 194.19 g·mol⁻¹) used as an isotopic standard and reagent in synthetic and analytical chemistry. It is supplied as a liquid, typically colorless to light yellow, and is intended for applications that require deuterium incorporation or accurate mass-based quantitation.
  • Deuterium-labeled (d15) for isotopic tracing and mass-spectrometry quantitation
  • Chemical formula C7D15Br; molecular weight 194.19 g·mol⁻¹
  • Supplied as a liquid, colorless to light yellow
  • Suitable as an internal standard for mass spectrometry
  • Useful in mechanistic studies, tracer experiments, and synthetic applications requiring deuterium