Learn More
Medchemexpress LLC 1-Bromoheptane-d15 | 98195-42-7 | MFCD01073970 | 99.2% | 194.19 g/mol | C7D15Br | 25 MG
SDP
Catalog No. 5000303531
Encompass_Preferred
1-Bromoheptane-d15 is the deuterium-labeled analogue of 1-bromoheptane used as a stable-isotope reagent in research and analytical chemistry. It provides a mass-shifted internal standard or tracer for mass spectrometry, quantitative analysis, and mechanistic studies, and is supplied in small pack sizes suitable for laboratory use.
  • Deuterium-labeled analogue of 1-bromoheptane.
  • Suitable as an internal standard for mass spectrometry and quantitative analysis.
  • Useful for tracer and mechanistic studies.
  • Available in small pack sizes (1-50 mg) for research use.
  • Supplied with product documentation, including SDS and data sheet.
Catalog No. 50-003-03531 Supplier Medchemexpress LLC Supplier No. 25MGHYW016577S
1-Bromoheptane-d15 is the deuterium-labeled analogue of 1-bromoheptane used as a stable-isotope reagent in research and analytical chemistry. It provides a mass-shifted internal standard or tracer for mass spectrometry, quantitative analysis, and mechanistic studies, and is supplied in small pack sizes suitable for laboratory use.
  • Deuterium-labeled analogue of 1-bromoheptane.
  • Suitable as an internal standard for mass spectrometry and quantitative analysis.
  • Useful for tracer and mechanistic studies.
  • Available in small pack sizes (1-50 mg) for research use.
  • Supplied with product documentation, including SDS and data sheet.