Learn More
Medchemexpress LLC 1-bromoheptane-d5 | 1219805-66-9 | 184.13 | C7H10D5Br | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW016577S210MG
Encompass_Preferred
1-Bromoheptane-d5 is the deuterium-labeled analogue of 1-bromoheptane used as an isotopic tracer and internal standard in analytical and synthetic chemistry. Supplied as a liquid with molecular formula C7H10D5Br and molecular weight 184.13, it is provided in mg-scale quantities suitable for mass spectrometry, quantitation, and deuterium-labeling studies.
  • Deuterium-labeled for use as an isotopic tracer and internal standard.
  • Suitable for mass spectrometry quantitation and method validation.
  • Provided in mg-scale quantities for analytical workflows.
  • Liquid form facilitates accurate spiking and sample preparation.
  • Molecular formula C7H10D5Br and CAS 1219805-66-9 for unambiguous identification.
Catalog No. 50-003-01121