Learn More
Medchemexpress LLC 1-bromoheptane-d5 | 1219805-66-9 | 95.0% | 184.13 g·mol⁻¹ | C7H10D5Br | 25 MG
SDP
Catalog No. 5000303032
Encompass_Preferred
1-Bromoheptane-d5 (CAS 1219805-66-9) is the deuterium-labeled analogue of 1-bromoheptane supplied as a research reagent and reference standard. It is used in analytical chemistry, mass spectrometry, tracer studies, and synthetic chemistry where isotopic labeling enables accurate quantification and tracking.
  • Deuterium-labeled (d5) to enable mass-differentiation in MS analyses.
  • Suitable as an internal standard for quantitative assays.
  • Liquid, ready for use in synthetic and analytical workflows.
  • Molecular formula C7H10D5Br and molecular weight 184.13 g·mol⁻¹.
  • Available in small pack sizes appropriate for research use.
Catalog No. 50-003-03032 Supplier Medchemexpress LLC Supplier No. 25MGHYW016577S2
1-Bromoheptane-d5 (CAS 1219805-66-9) is the deuterium-labeled analogue of 1-bromoheptane supplied as a research reagent and reference standard. It is used in analytical chemistry, mass spectrometry, tracer studies, and synthetic chemistry where isotopic labeling enables accurate quantification and tracking.
  • Deuterium-labeled (d5) to enable mass-differentiation in MS analyses.
  • Suitable as an internal standard for quantitative assays.
  • Liquid, ready for use in synthetic and analytical workflows.
  • Molecular formula C7H10D5Br and molecular weight 184.13 g·mol⁻¹.
  • Available in small pack sizes appropriate for research use.