Learn More
Medchemexpress LLC 1-bromoheptane-d5 | 1219805-66-9 | >98.0% | 184.13 | C7H10D5Br | 5 MG
SDP
Catalog No. 5000301122
Encompass_Preferred
1-Bromoheptane-d5 is a deuterium-labeled analogue of 1-bromoheptane supplied for research use as an isotope-labeled reagent. It is intended for analytical and synthetic chemistry applications, including use as an internal standard for mass spectrometry, tracer experiments, and mechanistic studies. Not for human use.
  • Deuterium-labeled analogue suitable as an internal standard for mass spectrometry.
  • High isotopic enrichment (approximately 99 atom % D) and chemical purity ≥98%.
  • Intended for tracer studies, mechanistic investigations, and isotope-labeling reactions.
  • Provided as a stable liquid for easy handling in small analytical quantities.
  • Recommended storage: -20°C (pure form); -80°C in solvent for extended preservation.
Catalog No. 50-003-01122 Supplier Medchemexpress LLC Supplier No. HYW016577S25MG
1-Bromoheptane-d5 is a deuterium-labeled analogue of 1-bromoheptane supplied for research use as an isotope-labeled reagent. It is intended for analytical and synthetic chemistry applications, including use as an internal standard for mass spectrometry, tracer experiments, and mechanistic studies. Not for human use.
  • Deuterium-labeled analogue suitable as an internal standard for mass spectrometry.
  • High isotopic enrichment (approximately 99 atom % D) and chemical purity ≥98%.
  • Intended for tracer studies, mechanistic investigations, and isotope-labeling reactions.
  • Provided as a stable liquid for easy handling in small analytical quantities.
  • Recommended storage: -20°C (pure form); -80°C in solvent for extended preservation.