Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-bromoheptane-d5 | 1219805-66-9 | >98.0% | 184.13 g/mol | C7H10D5Br | 50 MG
SDP

Catalog No. 5000301832
Encompass_Preferred

1-Bromoheptane-d5 is the deuterium-labeled analogue of 1-bromoheptane used as an isotope-labelled standard and reagent in analytical and synthetic applications. It contains five deuterium atoms, making it suitable as an internal standard for quantitative mass spectrometry and tracer studies. Store and handle under recommended low-temperature conditions to preserve integrity.

  • Deuterium-labeled (d5) isotopologue for mass spectrometry and tracer studies.
  • Suitable as an internal standard for quantitative GC-MS and LC-MS methods.
  • Molecular weight 184.13 g/mol and chemical formula C7H10D5Br.
  • Available in small-mass vials for laboratory use (for example, 50 mg).
  • Recommended storage: pure form -20°C; in solvent -80°C for long-term storage.

Catalog No. 50-003-01832 Supplier Medchemexpress LLC Supplier No. HYW016577S250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Bromoheptane-d5 is the deuterium-labeled analogue of 1-bromoheptane used as an isotope-labelled standard and reagent in analytical and synthetic applications. It contains five deuterium atoms, making it suitable as an internal standard for quantitative mass spectrometry and tracer studies. Store and handle under recommended low-temperature conditions to preserve integrity.

  • Deuterium-labeled (d5) isotopologue for mass spectrometry and tracer studies.
  • Suitable as an internal standard for quantitative GC-MS and LC-MS methods.
  • Molecular weight 184.13 g/mol and chemical formula C7H10D5Br.
  • Available in small-mass vials for laboratory use (for example, 50 mg).
  • Recommended storage: pure form -20°C; in solvent -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.