Learn More
Medchemexpress LLC 1-bromooctane | 111-83-1 | MFCD00000276 | 99.0% | 193.12 g/mol | C8H17Br | 1 KG
SDP
Catalog No. 5000300197
Encompass_Preferred
1-Bromooctane (n-octyl bromide) is a primary alkyl bromide commonly used as an alkylating reagent and as a substrate in nucleophilic substitution reactions. The terminal bromine acts as an effective leaving group, making the compound useful for SN2 and related synthetic transformations in organic chemistry and method development.
  • Primary alkyl bromide for alkylation reactions.
  • Terminal bromine provides an effective leaving group.
  • Useful substrate for nucleophilic substitution studies.
  • Applicable in organic synthesis and method development.
  • Compatible with common laboratory handling and storage procedures.
Catalog No. 50-003-00197 Supplier Medchemexpress LLC Supplier No. HYW0320151KG
1-Bromooctane (n-octyl bromide) is a primary alkyl bromide commonly used as an alkylating reagent and as a substrate in nucleophilic substitution reactions. The terminal bromine acts as an effective leaving group, making the compound useful for SN2 and related synthetic transformations in organic chemistry and method development.
  • Primary alkyl bromide for alkylation reactions.
  • Terminal bromine provides an effective leaving group.
  • Useful substrate for nucleophilic substitution studies.
  • Applicable in organic synthesis and method development.
  • Compatible with common laboratory handling and storage procedures.