Learn More
Medchemexpress LLC 1-bromooctane | 111-83-1 | MFCD00000276 | 99.0% | 193.12 g/mol | C8H17Br | 250 G
SDP
Catalog No. 5000302358
Encompass_Preferred
1-Bromooctane is a primary alkyl bromide (n-octyl bromide) used in organic synthesis as an alkylating reagent and as a substrate for nucleophilic substitution and cross-coupling reactions. It is a colorless liquid with CAS 111-83-1, molecular formula C8H17Br, and a molecular weight of 193.12 g/mol.
  • Primary alkyl bromide suitable for SN2 and alkylation reactions.
  • Terminal bromine provides a good leaving group for nucleophilic substitution.
  • Commonly used as a substrate in cross-coupling and chain-elongation chemistry.
  • Typically supplied with high purity (about 99.0%) for laboratory use.
  • Available in convenient laboratory pack sizes for synthesis workflows.
  • Colorless liquid, easy to handle under standard laboratory conditions.
Catalog No. 50-003-02358 Supplier Medchemexpress LLC Supplier No. 250GHYW032015
1-Bromooctane is a primary alkyl bromide (n-octyl bromide) used in organic synthesis as an alkylating reagent and as a substrate for nucleophilic substitution and cross-coupling reactions. It is a colorless liquid with CAS 111-83-1, molecular formula C8H17Br, and a molecular weight of 193.12 g/mol.
  • Primary alkyl bromide suitable for SN2 and alkylation reactions.
  • Terminal bromine provides a good leaving group for nucleophilic substitution.
  • Commonly used as a substrate in cross-coupling and chain-elongation chemistry.
  • Typically supplied with high purity (about 99.0%) for laboratory use.
  • Available in convenient laboratory pack sizes for synthesis workflows.
  • Colorless liquid, easy to handle under standard laboratory conditions.