Learn More
Medchemexpress LLC 1-Bromooctane | 111-83-1 | MFCD00000276 | 99.9% | 193.12 | C8H17Br | 500 G
SDP
Catalog No. 5000301727
Encompass_Preferred
1-Bromooctane is a primary alkyl bromide (CAS 111-83-1) commonly used as an electrophile in nucleophilic substitution and alkylation reactions. It functions as a building block in organic synthesis and is supplied for laboratory research applications.
  • Primary alkyl bromide suitable for SN2 reactions.
  • Useful electrophile for alkylation and substitution steps.
  • High purity for reliable reaction outcomes.
  • Available in multiple pack sizes for bench-scale synthesis.
  • Liquid at room temperature for easy handling and dosing.
Catalog No. 50-003-01727 Supplier Medchemexpress LLC Supplier No. HYW032015500G
1-Bromooctane is a primary alkyl bromide (CAS 111-83-1) commonly used as an electrophile in nucleophilic substitution and alkylation reactions. It functions as a building block in organic synthesis and is supplied for laboratory research applications.
  • Primary alkyl bromide suitable for SN2 reactions.
  • Useful electrophile for alkylation and substitution steps.
  • High purity for reliable reaction outcomes.
  • Available in multiple pack sizes for bench-scale synthesis.
  • Liquid at room temperature for easy handling and dosing.