Learn More
Medchemexpress LLC 1-bromooctane-d4 | 1219803-37-8 | 95.0% | 197.15 | C8H13D4Br | 1 MG
SDP
Catalog No. 5000299565
Encompass_Preferred
1-Bromooctane-d4 is the deuterium-labeled analogue of 1-bromooctane used as an isotope-labeled tracer and internal standard in analytical chemistry and mass spectrometry. It contains four deuterium atoms (d4), has molecular formula C8H13D4, and a molecular weight of 197.15 g·mol-1. Typical applications include quantitative analysis, structure elucidation, and mechanistic studies.
  • Deuterium-labeled internal standard for mass spectrometry quantification.
  • Useful tracer for metabolic and mechanistic studies.
  • Improves accuracy and precision in isotope-dilution assays.
  • Compatible with common organic solvents and GC/MS workflows.
  • Supplied in small reference quantities suitable for analytical use.
Catalog No. 50-002-99565 Supplier Medchemexpress LLC Supplier No. HYW032015S31MG
1-Bromooctane-d4 is the deuterium-labeled analogue of 1-bromooctane used as an isotope-labeled tracer and internal standard in analytical chemistry and mass spectrometry. It contains four deuterium atoms (d4), has molecular formula C8H13D4, and a molecular weight of 197.15 g·mol-1. Typical applications include quantitative analysis, structure elucidation, and mechanistic studies.
  • Deuterium-labeled internal standard for mass spectrometry quantification.
  • Useful tracer for metabolic and mechanistic studies.
  • Improves accuracy and precision in isotope-dilution assays.
  • Compatible with common organic solvents and GC/MS workflows.
  • Supplied in small reference quantities suitable for analytical use.