Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromopentane 98%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
110-53-2
C5H11Br
151.05
MFCD00000267
YZWKKMVJZFACSU-UHFFFAOYSA-N
n-amyl bromide,pentyl bromide,amyl bromide,pentane, 1-bromo,n-pentyl bromide,1-pentyl bromide,bromopentane,1-bromo-pentane,1-pentylbromide,unii-z2s4r599p0
8057
1-bromopentane
CCCCCBr
110-53-2
C5H11Br
151.05
MFCD00000267
YZWKKMVJZFACSU-UHFFFAOYSA-N
n-amyl bromide,pentyl bromide,amyl bromide,pentane, 1-bromo,n-pentyl bromide,1-pentyl bromide,bromopentane,1-bromo-pentane,1-pentylbromide,unii-z2s4r599p0
8057
1-bromopentane
CCCCCBr
Refractive Index 1.4430 to 1.446
Flash Point 31°C
Melting Point -95°C
Boiling Point 130°C
Density 1.2100g/mL
Specific Gravity 1.21
Color Colorless to Yellow
Quantity 1L
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)4Br
Beilstein 01, 131
Merck Index 15, 596
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,miscible with ether
Formula Weight 151.05
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1-Bromopentane, 98%
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-776-0

RTECSNumber : RZ9770000

TSCA : TSCA