Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Bromopropane

Catalog No. 501861078
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-186-1078 100 mL
50-186-1079 500 mL
50-186-1080 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-186-1078 Supplier Sigma Aldrich Supplier No. B78106100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Bromopropane
TRUSTED_SUSTAINABILITY

Specifications

CAS 106-94-5
Melting Point -110°C (lit.)
Density 1.354 g/mL (at 25°C (literature))
Boiling Point 71°C (lit.)
Molecular Formula C3H7Br
Refractive Index n20/D 1.434 (literature)
Linear Formula CH3CH2CH2Br
MDL Number MFCD00000254
Quantity 100 mL
Synonym Propyl bromide
Molecular Weight (g/mol) 122.99
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.