Learn More
Description
Specifications
Specifications
| Ignition Point | 914°F |
| CAS | 106-94-5 |
| Vapor Pressure | 146 mmHg (20°C) |
| Vapor Density | 4.3 (vs. air) |
| Density | 1.354 g/mL (at 25°C) |
| Linear Formula | CH3 CH2 CH2 Br |
| MDL Number | MFCD00000254 |
| Melting Point | -110°C |
| Boiling Point | 71°C |
| Synonym | Propyl bromide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.