Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromotetradecane, 98%

Catalog No. AC187381000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle

Catalog No. AC187381000

Supplier: Thermo Scientific Chemicals 187381000

Only null left
Add to Cart

CAS: 112-71-0 | C14H29Br | 277.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-71-0
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.28
MDL Number MFCD00000228
InChI Key KOFZTCSTGIWCQG-UHFFFAOYSA-N
Synonym myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo
PubChem CID 8208
IUPAC Name 1-bromotetradecane
SMILES CCCCCCCCCCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Yellow
Density 0.9300g/mL
Boiling Point 175.0°C to 178.0°C (20.0 mmHg)
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4590 to 1.4610
Linear Formula CH3(CH2)13Br
Quantity 100 g
Beilstein 01,II,136
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble
Formula Weight 277.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromotetradecane, 98%
Show More Show Less
EINECSNumber 203-999-3
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.