Learn More
Medchemexpress LLC 1-Bromotetradecane-d29 | 347841-80-9 | 98.4% | 306.46 | 1 MG
SDP
Catalog No. 5000454932
Encompass_Preferred
1-Bromotetradecane-d29 is the deuterium labeled 1-Bromotetradecane. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54932 Supplier Medchemexpress LLC Supplier No. HYW011591S1MG
1-Bromotetradecane-d29 is the deuterium labeled 1-Bromotetradecane. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.