Learn More
Medchemexpress LLC 1-Bromotetradecane-d29 | 347841-80-9 | 98.4% | 306.46 | 10 MG
SDP
Catalog No. 5000455147
Encompass_Preferred
1-Bromotetradecane-d29 is the deuterium labeled 1-Bromotetradecane. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-55147 Supplier Medchemexpress LLC Supplier No. HYW011591S10MG
1-Bromotetradecane-d29 is the deuterium labeled 1-Bromotetradecane. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.