Learn More
Medchemexpress LLC 1-Bromotetradecane-d3 | 347840-09-9 | 280.30 | 25 MG
SDP
Catalog No. 5000452707
Encompass_Preferred
1-Bromotetradecane-d3 is the deuterium labeled 1-Bromotetradecane. It is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes
  • Intended for research use only
Catalog No. 50-004-52707 Supplier Medchemexpress LLC Supplier No. 25MGHYW011591S
1-Bromotetradecane-d3 is the deuterium labeled 1-Bromotetradecane. It is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes
  • Intended for research use only