Learn More
AdipoGen 1-Bromoundecane
SDP
Catalog No. 502561674
Encompass_Preferred
Chemical. CAS 693-67-4. Formula C11H23Br. MW 235.2. Synthetic. Building block for synthesis. Reactant used in the preparation of Grignard reagent.
Catalog No. 50-256-1674 Supplier AdipoGen Supplier No. CDXB0116G250
Chemical. CAS 693-67-4. Formula C11H23Br. MW 235.2. Synthetic. Building block for synthesis. Reactant used in the preparation of Grignard reagent.