Learn More
Medchemexpress LLC 1-bromoundecane | 693-67-4 | 99.2% | 50 G
SDP
Catalog No. 5000232641
Encompass_Preferred
1-Bromoundecane is a primary alkyl bromide used as an organic synthesis reagent and intermediate in nucleophilic substitution and Grignard reactions. It is commonly applied as a building block in the preparation of long-chain alkyl derivatives and lipids.
  • Primary alkyl bromide suitable for nucleophilic substitution.
  • High purity for reliable synthetic performance.
  • Useful as a building block in lipid and long-chain alkyl syntheses.
  • Stable under recommended storage conditions; follow SDS for handling.
  • Packaged for laboratory use in a 50 g quantity.
Catalog No. 50-002-32641 Supplier Medchemexpress LLC Supplier No. HY12242650G
1-Bromoundecane is a primary alkyl bromide used as an organic synthesis reagent and intermediate in nucleophilic substitution and Grignard reactions. It is commonly applied as a building block in the preparation of long-chain alkyl derivatives and lipids.
  • Primary alkyl bromide suitable for nucleophilic substitution.
  • High purity for reliable synthetic performance.
  • Useful as a building block in lipid and long-chain alkyl syntheses.
  • Stable under recommended storage conditions; follow SDS for handling.
  • Packaged for laboratory use in a 50 g quantity.