Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-bromoundecane-d4 | 1219802-82-0 | 97.0% | 10 MG
SDP

Catalog No. 5000230853
Encompass_Preferred

1-Bromoundecane-d4 is the deuterium-labeled isotopologue of 1-bromoundecane used as a tracer and an internal standard in analytical chemistry and synthetic applications.

  • CAS number 1219802-82-0.
  • Molecular weight 239.23 g/mol.
  • Chemical formula C11H19D4Br.
  • Isotopic label d4 (four deuterium atoms).
  • Typical application as tracer or internal standard in mass spectrometry and analytical methods.
  • Supplied as a 10 mg packaged solid.

Catalog No. 50-002-30853 Supplier Medchemexpress LLC Supplier No. HY122426S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Bromoundecane-d4 is the deuterium-labeled isotopologue of 1-bromoundecane used as a tracer and an internal standard in analytical chemistry and synthetic applications.

  • CAS number 1219802-82-0.
  • Molecular weight 239.23 g/mol.
  • Chemical formula C11H19D4Br.
  • Isotopic label d4 (four deuterium atoms).
  • Typical application as tracer or internal standard in mass spectrometry and analytical methods.
  • Supplied as a 10 mg packaged solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.