Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-bromoundecane-d4 | 1219802-82-0 | 98.8% | 5 MG
SDP

Catalog No. 5000230714
Encompass_Preferred

1-Bromoundecane-d4 is the deuterium-labeled analogue of 1-bromoundecane supplied as an isotope-labeled standard for analytical and research use.

  • Isotope-labeled tracer and internal standard for analytical applications.
  • CAS 1219802-82-0.
  • Molecular formula C11H19D4Br.
  • Molecular weight 239.23 Da.
  • Purity 98.8% (as listed).
  • Available in small pack sizes such as 5 mg.

Catalog No. 50-002-30714 Supplier Medchemexpress LLC Supplier No. HY122426S25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Bromoundecane-d4 is the deuterium-labeled analogue of 1-bromoundecane supplied as an isotope-labeled standard for analytical and research use.

  • Isotope-labeled tracer and internal standard for analytical applications.
  • CAS 1219802-82-0.
  • Molecular formula C11H19D4Br.
  • Molecular weight 239.23 Da.
  • Purity 98.8% (as listed).
  • Available in small pack sizes such as 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.