Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-bromoundecane-d4 | 1219802-82-0 | 98.8% | 50 MG
SDP

Catalog No. 5000231291
Encompass_Preferred

Deuterium-labeled 1-bromoundecane (1-bromoundecane-d4) is supplied as a stable isotope standard for analytical and tracer applications. High purity and precise isotopic labeling make it suitable as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.

  • Deuterium-labeled isotopologue of 1-bromoundecane.
  • High purity: 98.8%.
  • Molecular formula C11H19D4Br.
  • Molecular weight 239.23 g/mol.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Available in 50 MG pack size.

Catalog No. 50-002-31291 Supplier Medchemexpress LLC Supplier No. HY122426S250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled 1-bromoundecane (1-bromoundecane-d4) is supplied as a stable isotope standard for analytical and tracer applications. High purity and precise isotopic labeling make it suitable as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.

  • Deuterium-labeled isotopologue of 1-bromoundecane.
  • High purity: 98.8%.
  • Molecular formula C11H19D4Br.
  • Molecular weight 239.23 g/mol.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Available in 50 MG pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.