Learn More
Medchemexpress LLC 1-butanaminium, N,N,N-tributyl-, iodide | 311-28-4 | MFCD00011636 | >99.9% | 369.37 g·mol^-1 | C16H36IN | 1mL
SDP
Catalog No. 5000268612
Encompass_Preferred
Tetrabutylammonium iodide is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Tetrabutylammonium iodide has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-68612 Supplier Medchemexpress LLC Supplier No. HYY079110MM/1ML
Tetrabutylammonium iodide is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Tetrabutylammonium iodide has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes