Learn More
Medchemexpress LLC 1-Butanaminium, N,N,N-tributyl-, perchlorate (1:1) | 1923-70-2 | MFCD00038722 | 98.0% | 341.91 g·mol⁻¹ | C16H36ClNO4 | 5 G
SDP
Catalog No. 5000283514
Encompass_Preferred
Tetrabutylammonium perchlorate is a quaternary ammonium salt commonly used as an electrolyte, phase-transfer reagent, and perchlorate anion source in organic synthesis and electrochemical applications. Supplied as a high-purity crystalline solid, it is suitable for laboratory research and analytical work.
  • High purity 98.0% suitable for research applications.
  • Functions as an electrolyte for electrochemical measurements.
  • Effective phase-transfer reagent for organic reactions.
  • Stable solid when stored sealed and away from moisture.
  • Available in lab-scale pack sizes for bench use.
Catalog No. 50-002-83514 Supplier Medchemexpress LLC Supplier No. HYW1276805G
Tetrabutylammonium perchlorate is a quaternary ammonium salt commonly used as an electrolyte, phase-transfer reagent, and perchlorate anion source in organic synthesis and electrochemical applications. Supplied as a high-purity crystalline solid, it is suitable for laboratory research and analytical work.
  • High purity 98.0% suitable for research applications.
  • Functions as an electrolyte for electrochemical measurements.
  • Effective phase-transfer reagent for organic reactions.
  • Stable solid when stored sealed and away from moisture.
  • Available in lab-scale pack sizes for bench use.