Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-butanaminium, N,N,N-tributyl-, trifluoromethanesulfonate (1:1) | 35895-70-6 | >98.0% | 391.53 g/mol | C17H36F3NO3S | 5 G
SDP

Catalog No. 5000210387
Encompass_Preferred

Tetrabutylammonium trifluoromethanesulfonate is a quaternary ammonium salt supplied as a biochemical assay reagent, commonly used as a phase-transfer catalyst and ion-pairing agent in organic synthesis and laboratory research.

  • Used as a phase-transfer catalyst in organic synthesis.
  • Functions as an ion-pairing agent in analytical and preparative procedures.
  • Typical purity greater than 98.0%.
  • Molecular weight 391.53 g/mol and formula C17H36F3NO3S.
  • Packaged as a 5 g sample for laboratory use.
  • Store at 4°C, sealed and protected from moisture.

Catalog No. 50-002-10387 Supplier Medchemexpress LLC Supplier No. HYW0425575G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrabutylammonium trifluoromethanesulfonate is a quaternary ammonium salt supplied as a biochemical assay reagent, commonly used as a phase-transfer catalyst and ion-pairing agent in organic synthesis and laboratory research.

  • Used as a phase-transfer catalyst in organic synthesis.
  • Functions as an ion-pairing agent in analytical and preparative procedures.
  • Typical purity greater than 98.0%.
  • Molecular weight 391.53 g/mol and formula C17H36F3NO3S.
  • Packaged as a 5 g sample for laboratory use.
  • Store at 4°C, sealed and protected from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.