Learn More
Medchemexpress LLC 1-Butanesulfonic acid, sodium salt | 2386-54-1 | MFCD00007540 | 98.0% | 160.17 g/mol | C4H9NaO3S | 1 G
SDP
Catalog No. 5000300154
Encompass_Preferred
Sodium butane-1-sulfonate (sodium 1-butanesulfonate) is an anionic sulfonate salt supplied as a white to off-white solid. It is used primarily as an ion-pairing reagent or mobile-phase additive in chromatographic separations, for example to improve separation of arsenic and selenium species. Typical purity is 98.0% and it is available in small laboratory pack sizes.
  • Anionic sulfonate suitable for ion-pairing in HPLC.
  • Improves separation of arsenic and selenium species.
  • White to off-white solid with stated purity of 98.0%.
  • Molecular weight 160.17 g/mol; formula C4H9NaO3S.
  • Supplied in small laboratory pack sizes for analytical use.
Catalog No. 50-003-00154 Supplier Medchemexpress LLC Supplier No. HYW0172241G
Sodium butane-1-sulfonate (sodium 1-butanesulfonate) is an anionic sulfonate salt supplied as a white to off-white solid. It is used primarily as an ion-pairing reagent or mobile-phase additive in chromatographic separations, for example to improve separation of arsenic and selenium species. Typical purity is 98.0% and it is available in small laboratory pack sizes.
  • Anionic sulfonate suitable for ion-pairing in HPLC.
  • Improves separation of arsenic and selenium species.
  • White to off-white solid with stated purity of 98.0%.
  • Molecular weight 160.17 g/mol; formula C4H9NaO3S.
  • Supplied in small laboratory pack sizes for analytical use.