Learn More
Medchemexpress LLC 1-Butanol, 2,2'-(1,2-ethanediyldiimino)bis-, hydrochloride (1:2), (2S,2'S)- | 1070-11-7 | 99.9% | 25 MG
SDP
Catalog No. 5000450955
Encompass_Preferred
Ethambutol dihydrochloride is an antimycobacterial compound that prevents cell wall formation by inhibiting arabinosyltransferase activity. This compound can also be used to establish animal models of hyperuricemia and optic neuropathy.
  • Targets bacterial and antibiotic pathways
  • Inhibits arabinosyltransferase activity to prevent cell wall formation
  • Can be used for modeling hyperuricemic disease in rats
  • Can be used for modeling optic neuropathy in rats
  • Appears as a white to off-white solid
  • Store at 4°C, sealed and away from moisture; in solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-50955 Supplier Medchemexpress LLC Supplier No. 25MGHYB0535AR
Ethambutol dihydrochloride is an antimycobacterial compound that prevents cell wall formation by inhibiting arabinosyltransferase activity. This compound can also be used to establish animal models of hyperuricemia and optic neuropathy.
  • Targets bacterial and antibiotic pathways
  • Inhibits arabinosyltransferase activity to prevent cell wall formation
  • Can be used for modeling hyperuricemic disease in rats
  • Can be used for modeling optic neuropathy in rats
  • Appears as a white to off-white solid
  • Store at 4°C, sealed and away from moisture; in solvent, store at -80°C for 6 months or -20°C for 1 month