Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ 1-Butanol
SCI_ED

Catalog No. S25209A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Reagent Grade
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25209A 500 mL Reagent Grade
S25209 100 mL Reagent Grade
S25209B 4 L Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25209A
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Color Colorless
Density 0.81g/mL
Quantity 500 mL
Packaging Reagent Grade
Assay Percent Range >99%
Linear Formula CH3(CH2)2CH2OH
Formula Weight 74.12
Grade Reagent
Chemical Name or Material 1-Butanol
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 3, III, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.