Learn More
U.S. Pharmacopeia 1-Butanol, 71-36-3, MFCD00002964, 3x1.2 mL
Catalog No. NC0645879
Encompass
Linear Formula: CH3(CH2)3OH, Molecular Weight: 74.12, bp: 116-118 °C, mp: −90 °C, Synonym: n-Butanol, Butyl alcohol.
Catalog No. NC0645879 Supplier U.S. Pharmacopeia Supplier No. 1081807
Linear Formula: CH3(CH2)3OH, Molecular Weight: 74.12, bp: 116-118 °C, mp: −90 °C, Synonym: n-Butanol, Butyl alcohol.