Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, 99.9%, Solstice

Catalog No. 60047079 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-079 1 L Stainless Steel Can
60-047-080 2.5 L Glass Bottle
60-047-084 6 x 1 L Glass Bottle
60-047-083 4 x 2.5 L Glass Bottle
60-047-081 20 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-047-079 Supplier Solstice Supplier No. 5379931L
Add to Cart
Add to Cart

≥99.9%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point ≤-90°C
Appearance Liquid
Density 0.81g/mL at 25°C
Quantity 1 L
Explosion Limit 11.2%
Ignition Point 649°C
Packaging Stainless Steel Can
Linear Formula CH3(CH2)3OH
UN Number UN1120
Beilstein 969148
Boiling Point 119°C
Molecular Weight (g/mol) 74.123
Formula Weight 74.12g/mol
Percent Purity 99%
Chemical Name or Material 1-Butanol
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
DOTInformation class 3 - PG 3 - Butanols
RTECSNumber 200-751-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.