Learn More
Sigma Aldrich Fine Chemicals Biosciences 1-Butanol 99.91L
Catalog No. 502898941
Encompass
1-Butanol is an industrial solvent that can be obtained either from petroleum feedstock or by biomass fermentation. Some of its characteristic properties include high energy content low vapor pressure and a better capability to combine with gasoline. Synthesis of di-n-butyl ether (DBE) in a fixed-bed flow reactor via dehydration of 1-butanol in the presence of³-alumina zeolites and Amberlyst ion-exchange resins has been investigated. Its thermophysical properties have been calculated. formation of fatty acid butyl esters (fABE) via transesterification of rapeseed and jatropha oil with 1-butanol in the presence of acid catalyst has been studied under varying conditions.
Catalog No. 50-289-8941 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5379931L
1-Butanol is an industrial solvent that can be obtained either from petroleum feedstock or by biomass fermentation. Some of its characteristic properties include high energy content low vapor pressure and a better capability to combine with gasoline. Synthesis of di-n-butyl ether (DBE) in a fixed-bed flow reactor via dehydration of 1-butanol in the presence of³-alumina zeolites and Amberlyst ion-exchange resins has been investigated. Its thermophysical properties have been calculated. formation of fatty acid butyl esters (fABE) via transesterification of rapeseed and jatropha oil with 1-butanol in the presence of acid catalyst has been studied under varying conditions.