Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, 99+%, Extra Dry, AcroSeal™

Catalog No. ac398961000
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398961000 100 mL Glass Bottle
AC398960010 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398961000 Supplier Thermo Scientific Chemicals Supplier No. 398961000
Only null left
Add to Cart

CAS: 71-36-3 | C4H10O | 74.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -89.0°C
Color Colorless
CAS Min % 99.0
CAS Max % 100.0
Density 0.8100g/mL
Quantity 100 mL
Flash Point 35°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3980 to 1.4000
Assay Percent Range 99+%
Residue after Evaporation 0.001% max.
Specific Gravity 0.81
Linear Formula CH3(CH2)3OH
Viscosity 2.95 mPa.s (20°C)
Water 0.005% max.
Merck Index 15, 1542
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: miscible with ethanol, ether and many other, organic solvents
Boiling Point 117.6°C
Formula Weight 74.12
Percent Purity ≥99%
Grade Extra Dry
Chemical Name or Material 1-Butanol, For spectroscopy
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.