Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, ACS Reagent, ≥99.4%, Solstice

Catalog No. 60046599 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-599 4 x 2.5 L Glass Bottle
60-046-598 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-599 Supplier Solstice Supplier No. 3604654X2.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.4%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -90°C
Density 0.81g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 0.112
Flash Point 35°C (95°F)
Ignition Point 649°F
Infrared Spectrum Conforms to structure
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.399
Linear Formula CH3(CH2)3OH
UN Number UN1120
Beilstein 969148
Boiling Point 116°C
Formula Weight 74.12g/mol
Percent Purity ≥99.4%
Chemical Name or Material 1-Butanol
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
DOTInformation class 3 - PG 3 - Butanols
RTECSNumber 200-751-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.