Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, Analytical Standard, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019394 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9394 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9394 Supplier MilliporeSigma Supelco Supplier No. 194225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butanol, alcohol with higher energy density, lower vapour pressure and low hygroscopicity. 1-Butanol is a potential fuel substitute to gasoline. It is also a feedstock used to produce chemicals. It is produced via two pathways, the synthetic 2-ketoacid pathway and CoA-dependent pathway. It can also be produced by fermentation of biomass-derived carbohydrates.

TRUSTED_SUSTAINABILITY

Specifications

CAS 71-36-3
Vapor Pressure 5 (4 mmHg) (at 20°C hPa)
Vapor Density 2.55 (vs. air)
Density 0.81 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3(CH2)3 OH
MDL Number MFCD00002964
Color Colorless
Melting Point -90°C
Boiling Point 116°C to 118°C
Synonym n-Butanol; Butyl alcohol
Quantity 5 mL
Formula Weight 74.12
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.